Learn More
Sino Biological Anti-IGF-II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106310T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse IGF-2/IGF-II, and purified by antigen affinity chromatography.
Catalog No. 50-240-9758