Learn More
Sino Biological Anti-IGF-II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50040T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IGF-2 / IGF-II (rM IGF-2 / IGF-II; Catalog#50040-M24E; P09535; Ala25-Glu91). IGF-2 / IGF-II specific IgG was purified by Mouse IGF-2 / IGF-II affinity chromatography.
Catalog No. 50-240-9021