Learn More
Sino Biological Anti-Igf2r Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101902T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse Igf2r, and purified by antigen affinity chromatography.
Catalog No. 50-241-1624