Learn More
Crystal Chem Inc Mouse IGFBP-3 ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80582
Encompass
The Mouse IGFBP-3 ELISA is used to quantify levels of mouse IGFBP-3 in plasma, serum, and cell culture using only a 5 µL sample volume. Highlights: Kits use only 5 µL sample, ideal for mouse applications. Dynamic range of 0.39 - 25 ng/mL. High sensitivity of 0.09 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7749