Learn More
Raybiotech Inc Mouse IGFBP-rp1/IGFBP-7 ELISA Kit
Supplier:  Raybiotech Inc ELMIGFBPRP11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse IGFBP-rp1.
Catalog No. 50-195-0024