Learn More
Abcam Mouse IGFBP2 ELISA Kit, 1 x 96 Tests
Catalog No. 502657614
Encompass_Preferred
Mouse IGFBP2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse IGFBP2 with a sensitivity of 103.6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7614 Supplier Abcam Supplier No. AB2076151X96TESTS
Mouse IGFBP2 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse IGFBP2 with a sensitivity of 103.6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement