Learn More
Sino Biological Anti-IGFBP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51124RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, human cell-derived, recombinant Human EGFR / ErbB / HER1. Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2995