Learn More
Sino Biological Mouse IGFBP6 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50460T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IGFBP6 / IBP6 (rM IGFBP6 / IBP6; Catalog#50460-M08H; NP_032370.2; Met1-Gly238). IGFBP6 / IBP6 specific IgG was purified by Mouse IGFBP6 / IBP6 affinity chromatography.
Catalog No. 50-237-3936