Learn More
ACROBiosystems HRP conjugated Anti-Human-IgG-Fc Antibody (6F11C8), mAb
Catalog No. 502331112
Encompass_Preferred
Immunoglobulins can be divided into five main classes/isotypes which are IgA, IgD, IgE, IgG, and IgM. IgG class identity is determined by class-specific sequences in the Fc region of the heavy chain. IgG antibody class are the most abundant immunoglobulins isotype in blood, lymph fluid, cerebrospinal fluid and peritoneal fluid. IgGs include four subclasses (IgG1, IgG2, IgG3, and IgG4). The IgG subclasses differ in their physical and chemical properties. Their distribution pattern is found to be age dependent and every subclass has a specific biological function.
Catalog No. 50-233-1112 Supplier ACROBiosystems Supplier No. IGGLY69100UG
Immunoglobulins can be divided into five main classes/isotypes which are IgA, IgD, IgE, IgG, and IgM. IgG class identity is determined by class-specific sequences in the Fc region of the heavy chain. IgG antibody class are the most abundant immunoglobulins isotype in blood, lymph fluid, cerebrospinal fluid and peritoneal fluid. IgGs include four subclasses (IgG1, IgG2, IgG3, and IgG4). The IgG subclasses differ in their physical and chemical properties. Their distribution pattern is found to be age dependent and every subclass has a specific biological function.