Learn More
Sigma Aldrich Fine Chemicals Biosciences Mouse IgG-Agarose Suspensi
Catalog No. 502900822
Encompass
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes namely IgG1 IgG2a IgG2b and IgG3. IgG1 regulates complement fixation in mice.
Catalog No. 50-290-0822 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A09192ML
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. Mouse IgGs have four distinct isotypes namely IgG1 IgG2a IgG2b and IgG3. IgG1 regulates complement fixation in mice.