Learn More
Chondrex Inc MOUSE IGG α COL2 ELISA, 1 KIT
Catalog No. 501526685
Encompass_Preferred
ELISA kit designed to assay mouse IgG antibodies against bovine type II collagen in mouse serum and plasma samples. This kit includes a 96-well ELISA plate with removable strips (10 bovine type II collagen coated strips and 4 standard coated strips), as well as all reagents necessary to complete the assay. Chromagen: TMB. Detection Range: 16 units/mL - 0.25 units/mL (100 units is approximately 0.1 µg IgG Ab/ml). Working Time: 4.25 hours. Ideal for quantifying immune response to bovine type II collagen used to immunize mice in the Collagen Induced Arthritis (CIA) model.
Catalog No. 50-152-6685 Supplier Chondrex Inc Supplier No. 2032T
ELISA kit designed to assay mouse IgG antibodies against bovine type II collagen in mouse serum and plasma samples. This kit includes a 96-well ELISA plate with removable strips (10 bovine type II collagen coated strips and 4 standard coated strips), as well as all reagents necessary to complete the assay. Chromagen: TMB. Detection Range: 16 units/mL - 0.25 units/mL (100 units is approximately 0.1 µg IgG Ab/ml). Working Time: 4.25 hours. Ideal for quantifying immune response to bovine type II collagen used to immunize mice in the Collagen Induced Arthritis (CIA) model.