Learn More
Chondrex Inc MOUSE IGG α COL2 ELISA, 1 KIT
Catalog No. 501526683
Encompass_Preferred
ELISA kit designed to assay mouse IgG antibodies against chick type II collagen in mouse serum and plasma samples. This kit includes a 96-well ELISA plate with removable strips (10 chick type II collagen coated strips and 4 standard coated strips), as well as all reagents necessary to complete the assay. Chromagen: TMB. Detection Range: 16 units/mL - 0.25 units/mL (100 units is approximately 0.1 µg IgG Ab/ml). Working Time: 4.25 hours. Ideal for quantifying immune response to chick type II collagen used to immunize mice in the Collagen Induced Arthritis (CIA) Model.
Catalog No. 50-152-6683 Supplier Chondrex Inc Supplier No. 2031T
Add to Cart
Add to Cart
ELISA kit designed to assay mouse IgG antibodies against chick type II collagen in mouse serum and plasma samples. This kit includes a 96-well ELISA plate with removable strips (10 chick type II collagen coated strips and 4 standard coated strips), as well as all reagents necessary to complete the assay. Chromagen: TMB. Detection Range: 16 units/mL - 0.25 units/mL (100 units is approximately 0.1 µg IgG Ab/ml). Working Time: 4.25 hours. Ideal for quantifying immune response to chick type II collagen used to immunize mice in the Collagen Induced Arthritis (CIA) Model.