Learn More
Genscript Corporation Mouse IgG control (Whole Molecule), Purified
Supplier:  Genscript Corporation A01007
Encompass_Preferred
GenScript Purified Mouse IgG (Whole Molecule) Control is highly purified from normal mouse serum through Protein A chromatography.GenScript Purified Mouse IgG (Whole Molecule) Control is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, immunobloting (Dot blot and Western blot), immunoprecipitation, immunodiffusion, and immunoelectrophoresis.It may also be used as antigen or ligand in immunochemical conjugation reaction.
Catalog No. 50-227-1944