Learn More
Mabtech Inc ELISpot Flex: Mouse IgG (ALP), For 4 plates, Detection of IgG (Total and Antigen-Specific), Flex format (without plates)
Supplier: Mabtech Inc 38252A
The Mabtech ELISpot Flex: Mouse IgG (ALP) kit is a versatile assay intended for the quantification of mouse B cells that secrete immunoglobulin G (IgG) antibodies. This kit allows researchers flexibility in experimental designs by providing the option to coat with antigen or use biotinylated antigen in detection, optimizing sensitivity and spot quality. Key reagents included facilitate the effective pre-stimulation of memory B cells and are essential for assay performance.
- Detects both total and antigen-specific mouse IgG-secreting B cells
- Includes capture and detection antibodies, streptavidin-ALP conjugate, and activators
- Adaptable for various experimental designs
- Contains essential buffers for stability and effectiveness
- Shelf life of at least 18 months, ensuring long-term usability
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.