Learn More
ACROBiosystems Mouse IgG Fc 1mg
Catalog No. 502534623
Encompass_Preferred
Monoclonal Anti-Human-IgG-Fc Antibody Mouse IgG1 is a chimeric monoclonal antibody recombinantly expressed from CHO cells which combines the variable region of a mouse monoclonal antibody with mouse IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with IgG Fc.
Catalog No. 50-253-4623 Supplier ACROBiosystems Supplier No. IGGS3071MG
Monoclonal Anti-Human-IgG-Fc Antibody Mouse IgG1 is a chimeric monoclonal antibody recombinantly expressed from CHO cells which combines the variable region of a mouse monoclonal antibody with mouse IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with IgG Fc.