Learn More
Rockland Immunochemicals Mouse IgG (H&L) Secondary Antibody 2 mg
SDP
Catalog No. 501943905
Encompass_Preferred
Anti-Mouse IgG Antibody generated in rabbit detects reactivity to Mouse IgG heavy and light chains. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3905 Supplier Rockland Immunochemicals Supplier No. 6104102
Anti-Mouse IgG Antibody generated in rabbit detects reactivity to Mouse IgG heavy and light chains. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.