Learn More
Sino Biological Mouse IgG Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786743
Encompass_Preferred
The Human SH2D1A ELISA Pair Set is for the quantitative determination of Mouse IgG.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IgG coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IgG present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IgG polyclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IgG present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86743 Supplier Sino Biological Supplier No. MGP5
The Human SH2D1A ELISA Pair Set is for the quantitative determination of Mouse IgG.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IgG coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IgG present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IgG polyclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IgG present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.