Learn More
Sino Biological Rabbit Anti-Mouse IgG Antibody (Heavy and Light chains), Rabbit PAb
Catalog No. 502408790
Encompass_Preferred
Produced in rabbits immunized with purified mouse IgG (heavy and light chains). This polyclonal antibody was purified by Protein A affinity chromatography.
Catalog No. 50-240-8790 Supplier Sino Biological Supplier No. 50323RP01100
Produced in rabbits immunized with purified mouse IgG (heavy and light chains). This polyclonal antibody was purified by Protein A affinity chromatography.