Learn More
Chondrex Inc MOUSE IGG1 ANTI-OVA ASSAY 1 KT
Supplier:  Chondrex Inc 3013
Encompass_Preferred
ELISA kit for assaying mouse anti-ovalbumin (OVA) IgG1 antibodies found in mouse serum and plasma samples. This kit includes one 96-well ELISA plate with removable strips, as well as all reagents necessary to complete the assay. Ideal for quantifying the immune response to ovalbumin used to induce allergy models in mice. Standard Range: 12.5 ng/ml - 0.2 ng/ml. Working Time: 4.5 hours.
Catalog No. 50-152-6742