Learn More
Sino Biological Mouse IgG1, kappa isotype control (FITC)
Supplier:  Sino Biological IC003M063F25
Encompass_Preferred
The isotype has unknown specificity. The isotype of this antibody is mouse IgG1, ?. This antibody was chosen as an isotype control after screening on a variety of cells and tissues.
Catalog No. 50-240-8683