Learn More
AdipoGen Mouse IgG1 Isotype Ctrl
SDP
Supplier:  AdipoGen AG35B0003C050
Encompass_Preferred
Monoclonal Antibody. Isotype Mouse IgG1. Clone 15H6. Applications FACS, ICC, IHC, IP. Liquid. In PBS containing 0.02% sodium azide. Negative control in assays where mouse IgG1 antibodies are used.
Catalog No. 50-256-3094