Learn More
Rockland Immunochemicals MOUSE IgG1 Lambda (λ) isotype control - 010-001-331
SDP
Supplier:  Rockland Immunochemicals 010001331
Encompass
Mouse isotype controls are used in flow cytometry, western blot and ELISA and differentiate between immunoglobulin classes and subclasses. Isotype controls allow for the genetic variations or differences in the constant regions of the heavy and light chains. In mouse there are six relevant heavy chain isotypes and two light chain isotypes: heavy chain alpha - IgA, gamma - IgG 1, 2a, 2b, 3 and μ - IgM, light chain kappa and lambda. IgG1 is the most abundant of the four IgG subclasses. This isotype control possesses lambda light chains.
Catalog No. NC1277827