Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG2A Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB0033100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB0033100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB0033100 Supplier R&D Systems Supplier No. MAB0033100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Flow Cytometry∣Western Blot.

Specifications

Antigen IgG2a
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2214A
Conjugate Unconjugated
Dilution Western Blot 0.5μg/mL, Flow Cytometry 0.25μg/106 cells
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Host Species Rabbit
Immunogen Mouse IgG
Quantity 100 μg
Primary or Secondary Secondary
Test Specificity Detects mouse IgG 2A in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with mouse IgG 1 mouse IgG 2B and mouse IgG 3 was observed.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.