Learn More
Genscript Corporation Mouse IgG2a control (Whole Molecule), Purified
Supplier:  Genscript Corporation A02000
Encompass_Preferred
Mouse IgG2a control (Whole Molecule), Purified is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, FACS, ICC immunoblotting, Dot blot, Western blot, immunoprecipitation, immunodiffusion, and immunoelectrophoresis. It may also be used as antigen or ligand in immunochemical conjugation reaction.GenScript Mouse IgG2a control (Whole Molecule), Purified is highly purified from cell culture in vitro under conditions free from animal-derived components through Protein A affinity column.
Catalog No. 50-227-2016