Learn More
Abcam Mouse IgG2a ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1330461X96TESTS
Encompass_Preferred
Mouse IgG2a ELISA Kit ab133046 is used to quantify Mouse IgG2a with a sensitivity of 47.3 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2111991