Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG2a Uncoated ELISA Kit with Plates

Catalog No. 50246300
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-300 2 x 96 Tests
50-246-303 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-246-300 Supplier Invitrogen™ Supplier No. 885042022
Add to Cart
Add to Cart

ELISA

The Mouse Immunoglobulin G2a (IgG2a) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IgG2a. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.90-250 ng/mL
Assay Sensitivity 3.90 ng/mL
Content And Storage Capture Antibody Concentrate
Detection Antibody Concentrate
Mouse IgG2a isotype control (standard), lyophilized, 500 ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Tetramethylbenzidine (TMB) Substrate Solution
96-well plates included with product Cat. Nos. ending in suffixes -22, -76, -86
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit with Plates
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2;
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye HRP
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 10 μL, Serum 10 μL, Supernatant 50 μL
Storage Requirements 2-8°C
Synonym Immunoglobulin G2a
Target Mouse IgG2a
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
What type of plates are included in the Ready-SET-Go! ELISA sets?

Corning Costar 96-well plates are included with Ready-SET-Go! ELISA catalog numbers ending in suffixes -22, -86 and -76. These solid 96-well plates are optional with the 10 and 20 plate kit sizes.

What is the sensitivity of your Ready-Set-Go! ELISA Sets?

The sensitivity ranges of our Ready-SET-Go! ELISA sets vary depending on the cytokine/protein being measured. Please refer to the product datasheet for set specific information.

What is the expiration date for your Ready-Set-Go! ELISA Sets?

The majority of our Ready-Set-Go! ELISA Sets are guaranteed for one year from the date of shipment. However, we do have several sets that are guaranteed for 6 months upon date of receipt. Please refer to the Certificate of Analysis for shelf-life information.

What are the storage conditions for your Ready-Set-Go! ELISA Sets?

All of the reagents are to be stored at 4 degrees C upon receipt, except for the recombinant standards. The recombinant standards MUST be stored at or below -80 degrees C to ensure stability and performance.

What do Ready-SET-Go! ELISA Sets contain?

Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant cytokine for generating standard curve and calibrating samples
10X Coating Buffer
5X ELISA/ELISPOT Diluent
Detection enzyme: pre-titrated Avidin-HRP
Substrate Solution: 1X TMB (Tetramethylbenzidine) Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.