Learn More
Medchemexpress LLC Mouse IgG2a Fc Isot 10mg | 10mg
SDP
Catalog No. 5000325353
Encompass_Preferred
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa
Catalog No. 50-003-25353 Supplier Medchemexpress LLC Supplier No. HYP99000510MG
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa