Learn More
Medchemexpress LLC Mouse IgG2a Fc Isot 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9900051MG
Encompass_Preferred
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa
Catalog No. 50-003-24787