Learn More
Medchemexpress LLC Mouse IgG2a Fc Isot 25mg | 25mg
SDP
Catalog No. 5000325087
Encompass_Preferred
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa
Catalog No. 50-003-25087 Supplier Medchemexpress LLC Supplier No. HYP99000525MG
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa