Learn More
Medchemexpress LLC Mouse IgG2a Fc Isot 5mg | 5mg
SDP
Catalog No. 5000324738
Encompass_Preferred
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa
Catalog No. 50-003-24738 Supplier Medchemexpress LLC Supplier No. HYP9900055MG
Mouse IgG2a Fc Isotype Control is a Fc fragment of mouse IgG2a only and does not contain the Fab fragments The molecular mass is about 52 kDa