Learn More
ACROBiosystems ACROBiosystems Mouse IgG2a Fc Protein, Tag Free
Supplier:  ACROBiosystems IGAM5207200UG
Encompass_Preferred
Recombinant Protein;200UG;Mouse IgG2a Fc, Tag Free (IGA-M5207) is expressed from human 293 cells (HEK293). It contains AA Glu 98 - Lys 330 (Accession # CAC20702).;This protein carries no "tag". The protein has a calculated MW of 26.4 kDa. As a result of glycosylation, the protein migrates as 30-35 kDa under reducing (R) condition, and 55-60 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-7492