Learn More
American Research Products Inc Mouse IgG2a Fc Silenced
Catalog No. 5000305439
Encompass
Mouse IgG2a Fc Silenced (D265A) Isotype Control Purified No Carrier Protein is a isotype control produced in mouse Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-05439 Supplier American Research Products Inc Supplier No. IVMB055325MG
Mouse IgG2a Fc Silenced (D265A) Isotype Control Purified No Carrier Protein is a isotype control produced in mouse Available in 1mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles