Learn More
Rockland Immunochemicals Mouse IgG2a (Gamma 2a chain) Antibody Peroxidase Conjugated 1 mg
SDP
Catalog No. 501943909
Encompass_Preferred
Anti-Mouse IgG2a heavy chain antibody generated in rabbit detects specifically Mouse IgG2a heavy chain. This secondary antibody anti-Mouse is ideal for investigators who routinely perform titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3909 Supplier Rockland Immunochemicals Supplier No. 6104341
Anti-Mouse IgG2a heavy chain antibody generated in rabbit detects specifically Mouse IgG2a heavy chain. This secondary antibody anti-Mouse is ideal for investigators who routinely perform titration assays western-blot immunoprecipitation and more generally immunoassays.