Learn More
ACROBiosystems ACROBiosystems Mouse IgG2b Fc Protein, Tag Free (MALS verified)
Supplier:  ACROBiosystems IGBM52031MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse IgG2b Fc, Tag Free (IGB-M5203) is expressed from human 293 cells (HEK293). It contains AA Glu 97 - Lys 335 (Accession # P01867-2).;This protein carries no "tag". The protein has a calculated MW of 26.7 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7402