Learn More
AdipoGen Mouse IgG2b Isotype Ctrl
SDP
Supplier:  AdipoGen AG35B0005C050
Encompass_Preferred
Monoclonal Antibody. Isotype Mouse IgG2bkappa. Clone A-1. Applications FACS, ICC, IHC, IP. Liquid. In PBS containing 0.02% sodium azide. Negative control in assays where mouse IgG2b antibodies are used.
Catalog No. 50-256-3088