Learn More
CAPRICO BIOTECHNOLOGIES INC Caprico Biotechnologies 1120195 CD4 iFluor and CD8 iFluor Antibody Panel
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4024195
Encompass_Preferred
The Caprico Biotechnologies 1120195 CD4 iFluor is a monoclonal antibody that targets human CD4. It is conjugated to iFluor 488 and is suitable for flow cytometry, immunofluorescence microscopy, and immunohistochemistry. This antibody is designed for the identification and enumeration of CD4+ T lymphocytes. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each test contains 100 μL of antibody. The antibody should be stored at 2-8°C and is stable for 12 months from date of receipt. This product is for research use only.
Catalog No. 50-227-9341