Learn More
CAPRICO BIOTECHNOLOGIES INC IgG2b (K) mFluor 540, MPC11 ASR
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4024165
Encompass_Preferred
The CapricoBio IGG2B K-MFluor 540 MPC11 ASR is a monoclonal antibody conjugated to MFluor 540 fluorescent dye. It is an anti-mouse immunoglobulin G2b (IgG2b) antibody produced in rat and has an excitation max of 540nm and emission max of 565nm. This antibody is pre-diluted and ready to use in flow cytometry applications for labeling and detecting mouse IgG2b antibodies. It is optimized for use with CapricoBio Kits and Reagents.
Catalog No. 50-227-9340