Learn More
Sino Biological Recombinant Mouse IgG2b Protein, HPLC-verified 1
Supplier:  Sino Biological 51095MNAH1
Encompass_Preferred
A DNA sequence encoding the Mouse IgG2b Fc region (P01867-2) (Glu 97-Lys 335) was expressed and purified.
Catalog No. 50-224-6852