Learn More
Sino Biological Recombinant Mouse IgG3 Protein 1
Supplier:  Sino Biological 51096MNAH1
Encompass_Preferred
A DNA sequence encoding the Mouse IgG3 Fc region (P03987-2) (Glu 97-Lys 329) was expressed and purified.
Catalog No. 50-224-6853