Learn More
Sino Biological Recombinant Mouse IgG2a Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 51094MNAHB100
Encompass_Preferred
A DNA sequence encoding the mouse IgG2a Fc region (P01863) (Glu98-Lys330) was expressed and purified.The purified protein was biotinylated in vitro.
Catalog No. 50-255-3013