Learn More
Sino Biological Rabbit Anti-Mouse IgM / Immunoglobulin M Secondary Antibody (HRP), Rabbit MAb
Catalog No. 502408253
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified mouse IgM protein, and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8253 Supplier Sino Biological Supplier No. 68045R118H100
This antibody was obtained from a rabbit immunized with purified mouse IgM protein, and conjugated with horseradish-peroxidase (HRP).