Learn More
BIOHIPPO Mouse Igk Microsample
SDP
Catalog No. 5000315209
Encompass
Mouse Igk Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Igk in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 ng/mL with a sensitivity of 35 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Igk Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Igk Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Igk biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15209 Supplier BIOHIPPO Supplier No. BHE15210759
Mouse Igk Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Igk in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 ng/mL with a sensitivity of 35 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Igk Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Igk Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Igk biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color