Learn More
Raybiotech Inc Mouse IgM ELISA Kit
Supplier:  Raybiotech Inc ELMIGM1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 20 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse heavy chain of IgM. Other species not determined.
Catalog No. 50-195-0029