Learn More
Abcam Mouse IgM ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1330471X96TESTS
Encompass_Preferred
Mouse IgM ELISA Kit ab133047 is used to quantify Mouse IgM with a sensitivity of 0.6 ng/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2241175