Learn More
Rockland Immunochemicals MOUSE IgM whole molecule - 010-0107
SDP
Supplier:  Rockland Immunochemicals 0100107
Encompass
Mouse IgM, or mouse Immunoglobulin M, purified protein is a basic antibody that is produced by B cells. Mouse IgM is by far the physically largest antibody in the human circulatory system. Mouse IgM is the first antibody to appear in response to initial exposure to antigen. IgM is predominantly produced in the spleen. Mouse IgM is formed from covalently linking 5 immunoglobulins together, the approixmate molecular weight of IgM is 900kDa and possesses 10 binding sites.
Catalog No. 50-105-7358