Learn More
Kingfisher Biotech Inc Mouse IL-1 alpha Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0905M025
Encompass_Preferred
The Mouse IL-1 alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse IL-1 alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Mouse IL-1 alpha yeast-derived recombinant protein can be purchased in multiple sizes. Mouse IL-1 alpha Specifications: (Molecular Weight: 18.0 kDa) (Amino Acid Sequence: SAPYTYQSDL RYKLMKLVRQ KFVMNDSLNQ TIYQDVDKHY LSTTWLNDLQ QEVKFDMYAY SSGGDDSKYP VTLKISDSQL FVSAQGEDQP VLLKELPETP KLITGSETDL IFFWKSINSK NYFTSAAYPE LFIATKEQSR VHLARGLPSM TDFQIS (156)) (Gene ID: 16175). For research use only. Made in the USA
Catalog No. 50-241-4022