Learn More
Sino Biological Mouse IL-1 alpha/IL1A Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK501145
The Mouse IL-1 alpha / IL-1F1 / IL1A ELISA Pair Set is for the quantitative determination of Mouse IL-1 alpha / IL-1F1 / IL1A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IL-1 alpha / IL-1F1 / IL1A coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IL-1 alpha / IL-1F1 / IL1A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse IL-1 alpha / IL-1F1 / IL1A polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IL-1 alpha / IL-1F1 / IL1A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.