Learn More
Abcam Mouse IL-1 beta ELISA Kit, Fluorescent, 1 x 96 Tests
Supplier:  Abcam AB2294401X96TESTS
Encompass_Preferred
Mouse IL-1 beta ELISA Kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Mouse IL-1 beta with a sensitivity of 0.2 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1970331