Learn More
ACROBiosystems Mouse IL-1 RAcP, His Tag (ILP-M52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Accession # Q61730-1).
Supplier:  ACROBiosystems ILPM52H5100UG
Encompass_Preferred
Interleukin-1 receptor accessory protein (IL1RAP or IL-1RAcP) is also known as Interleukin-1 receptor 3 (IL-1R-3 or IL-1R3), C3orf13, which belongs to the interleukin-1 receptor family. IL1RAP contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL1RAP is detected in liver, skin, placenta, thymus and lung. IL-1R3 / IL1RAP is coreceptor with IL1R1, associates with IL1R1 bound to IL1B to form the high affinity interleukin-1 receptor complex which mediates interleukin-1-dependent activation of NF-kappa-B and other pathways.
Catalog No. 50-223-0944