Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL1RAPL1 (P59823) VersaClone cDNA

Catalog No. RDC2457
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2457 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2457 Supplier R&D Systems™ Supplier No. RDC2457
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Mouse IL-1 RAPL1/IL-1 R8 (P59823) VersaClone cDNA clone contains the complete ORF for IL-1 RAPL1/IL-1 R8, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The IL-1 RAPL1/IL-1 R8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided

Specifications

Gene mIL-1RAPL1/IL-1R8
Accession Number P59823
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 11141
Gene Alias IL1R8IL1RAPL, IL-1R9, IL-1 RAPL1, IL-1-RAPL-1, IL-1 RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MR x 10, MR x 21, MR x 34, oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2, TIGIRR-2 mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1
Content And Storage Store the unopened product at -20 to -80°C
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.